missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxyphenol 98.0+%, TCI America™

Catalog No. E021625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E021625G 25 g
E0216500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E021625G Supplier TCI America Supplier No. E021625G

Chemical Identifiers

CAS 622-62-8
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00002334
InChI Key LKVFCSWBKOVHAH-UHFFFAOYSA-N
Synonym phenol, 4-ethoxy, p-ethoxyphenol, hydroquinone monoethyl ether, 4-ethyloxyphenol, p-hydroxyphenetole, phenol, p-ethoxy, 4-ethoxy-phenol, 1-ethoxy-4-hydroxybenzene, para-ethoxyphenol, ethoxyphenol, p
PubChem CID 12150
IUPAC Name 4-ethoxyphenol
SMILES CCOC1=CC=C(C=C1)O

Specifications

Melting Point 65°C
Boiling Point 121°C
Quantity 25 g
Formula Weight 138.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Ethoxyphenol
EINECSNumber (3)-0568
RTECSNumber SL3790000
TSCA Yes