accessibility menu, dialog, popup

UserName

CH3OOH

CAS RN 622-62-8

IUPAC Name: 4-ethoxyphenol
Synonyms: P-ethoxyphenol
Molecular Weight (g/mol): 138.17
Molecular Formula: C8H10O2
InChi Key: LKVFCSWBKOVHAH-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(O)C=C1
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)