missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxyphenol, 99%

Catalog No. AC409670250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC409670250 25 g Glass bottle
AC409671000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC409670250 Supplier Thermo Scientific Chemicals Supplier No. 409670250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 622-62-8 | C8H10O2 | 138.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 622-62-8
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00002334
InChI Key LKVFCSWBKOVHAH-UHFFFAOYSA-N
Synonym phenol, 4-ethoxy, p-ethoxyphenol, hydroquinone monoethyl ether, 4-ethyloxyphenol, p-hydroxyphenetole, phenol, p-ethoxy, 4-ethoxy-phenol, 1-ethoxy-4-hydroxybenzene, para-ethoxyphenol, ethoxyphenol, p
PubChem CID 12150
IUPAC Name 4-ethoxyphenol
SMILES CCOC1=CC=C(C=C1)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 64°C to 67°C
Color Beige
Boiling Point 131°C (9.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: slightly soluble. Other solubilities: very soluble in ethanol,ethyl ether,soluble in chloroform
Formula Weight 138.17
Percent Purity 99%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 4-Ethoxyphenol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-748-1

RUO – Research Use Only