missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Ethoxybenzonitrile 98.0+%, TCI America™
$38.51 - $117.44
Chemical Identifiers
| CAS | 25117-74-2 |
|---|---|
| Molecular Formula | C9H9NO |
| Molecular Weight (g/mol) | 147.177 |
| MDL Number | MFCD00001819 |
| InChI Key | PJRLUGQMEZZDIY-UHFFFAOYSA-N |
| Synonym | benzonitrile, 4-ethoxy, p-ethoxycyanobenzene, p-ethoxybenzonitrile, 4-ethoxybenzoic acid nitrile, benzonitrile, p-ethoxy, 4-ethoxybenzenecarbonitrile, 4-ethoxy-benzonitrile, 4-ethyloxybenzonitrile, acmc-1cis8 |
| PubChem CID | 141176 |
| IUPAC Name | 4-ethoxybenzonitrile |
| SMILES | CCOC1=CC=C(C=C1)C#N |
Chemical Identifiers
| 25117-74-2 | |
| 147.177 | |
| PJRLUGQMEZZDIY-UHFFFAOYSA-N | |
| 141176 | |
| CCOC1=CC=C(C=C1)C#N |
| C9H9NO | |
| MFCD00001819 | |
| benzonitrile, 4-ethoxy, p-ethoxycyanobenzene, p-ethoxybenzonitrile, 4-ethoxybenzoic acid nitrile, benzonitrile, p-ethoxy, 4-ethoxybenzenecarbonitrile, 4-ethoxy-benzonitrile, 4-ethyloxybenzonitrile, acmc-1cis8 | |
| 4-ethoxybenzonitrile |
Specifications
| 25117-74-2 | |
| White-Yellow | |
| C9H9NO | |
| 5 g | |
| benzonitrile, 4-ethoxy, p-ethoxycyanobenzene, p-ethoxybenzonitrile, 4-ethoxybenzoic acid nitrile, benzonitrile, p-ethoxy, 4-ethoxybenzenecarbonitrile, 4-ethoxy-benzonitrile, 4-ethyloxybenzonitrile, acmc-1cis8 | |
| CCOC1=CC=C(C=C1)C#N | |
| 147.177 | |
| 147.18 | |
| Crystalline Powder |
| 62°C | |
| 145°C | |
| MFCD00001819 | |
| 3439 | |
| PJRLUGQMEZZDIY-UHFFFAOYSA-N | |
| 4-ethoxybenzonitrile | |
| 141176 | |
| ≥98.0% (GC) | |
| 4-Ethoxybenzonitrile |
Safety and Handling
TSCA : No