accessibility menu, dialog, popup

UserName

CAS RN 25117-74-2

CH3ON

CAS RN 25117-74-2

IUPAC Name: 4-ethoxybenzonitrile
Synonyms: benzonitrile, 4-ethoxy-
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: PJRLUGQMEZZDIY-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(C=C1)C#N
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)