missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxybenzonitrile 98.0+%, TCI America™

Catalog No. E09815G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09815G 5 g
E098125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09815G Supplier TCI America Supplier No. E09815G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 25117-74-2
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00001819
InChI Key PJRLUGQMEZZDIY-UHFFFAOYSA-N
Synonym benzonitrile, 4-ethoxy, p-ethoxycyanobenzene, p-ethoxybenzonitrile, 4-ethoxybenzoic acid nitrile, benzonitrile, p-ethoxy, 4-ethoxybenzenecarbonitrile, 4-ethoxy-benzonitrile, 4-ethyloxybenzonitrile, acmc-1cis8
PubChem CID 141176
IUPAC Name 4-ethoxybenzonitrile
SMILES CCOC1=CC=C(C=C1)C#N

Specifications

Melting Point 62°C
Color White-Yellow
Boiling Point 145°C
Quantity 5 g
UN Number 3439
Formula Weight 147.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Ethoxybenzonitrile