missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxybenzoic acid, 98+%

Catalog No. AAA1230914
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1230914 25 g
AAA1230922 100 g
AAA1230936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1230914 Supplier Thermo Scientific Chemicals Supplier No. A1230914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 619-86-3 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 619-86-3
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00002545
InChI Key SHSGDXCJYVZFTP-UHFFFAOYSA-N
Synonym p-ethoxybenzoic acid, benzoic acid, 4-ethoxy, benzoic acid, p-ethoxy, 4-ethoxy-benzoic acid, unii-tzr23xeq7w, tzr23xeq7w, 4-ethoxybenzoicacid, 4ethoxybenzoic acid, pubchem2686, para-ethoxybenzoic acid
PubChem CID 12093
IUPAC Name 4-ethoxybenzoic acid
SMILES CCOC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 197°C to 199°C
Quantity 25 g
Beilstein 2207349
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material 4-Ethoxybenzoic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 210-616-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only