accessibility menu, dialog, popup

UserName

CH3OOHO

CAS RN 619-86-3

IUPAC Name: 4-ethoxybenzoic acid
Synonyms: P-ethoxybenzoic acid
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: SHSGDXCJYVZFTP-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(C=C1)C(O)=O
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)