missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxy-3-nitropyridine 98.0+%, TCI America™

Catalog No. E09421G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09421G 1 g
E09425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09421G Supplier TCI America Supplier No. E09421G
Only null left

Chemical Identifiers

CAS 1796-84-5
Molecular Formula C7H8N2O3
Molecular Weight (g/mol) 168.152
MDL Number MFCD00234959
InChI Key ABOSMHRLVUWEMT-UHFFFAOYSA-N
PubChem CID 356305
IUPAC Name 4-ethoxy-3-nitropyridine
SMILES CCOC1=C(C=NC=C1)[N+](=O)[O-]

Specifications

Melting Point 49°C
Color Yellow
Quantity 1 g
Formula Weight 168.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Ethoxy-3-nitropyridine