accessibility menu, dialog, popup

UserName

CH3ONNOO

CAS RN 1796-84-5

IUPAC Name: 4-ethoxy-3-nitropyridine
Synonyms: 4-ethoxy-3-nitropyridine
Molecular Weight (g/mol): 168.15
Molecular Formula: C7H8N2O3
InChi Key: ABOSMHRLVUWEMT-UHFFFAOYSA-N
SMILES: CCOC1=C(C=NC=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results