missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminobenzophenone, 98%

Catalog No. AAB2038906
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2038906 5 g
AAB2038914 25 g
AAB2038922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2038906 Supplier Thermo Scientific Chemicals Supplier No. B2038906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 530-44-9 | C15H15NO | 225.291 g/mol

4-(Dimethylamino)benzophenone is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Dimethylamino)benzophenone is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 530-44-9
Molecular Formula C15H15NO
Molecular Weight (g/mol) 225.291
MDL Number MFCD00008311
InChI Key BEUGBYXJXMVRFO-UHFFFAOYSA-N
Synonym 4-dimethylamino benzophenone, 4-dimethylaminobenzophenone, p-dimethylaminobenzophenone, p-benzoyl-n,n-dimethylaniline, 4-n,n-dimethylaminobenzophenone, p-dimethylamino benzophenone, 4-dimethylamino phenyl phenyl methanone, 4-benzoyl-n,n-dimethylaniline, methanone, 4-dimethylamino phenyl phenyl, benzophenone, 4-dimethylamino
PubChem CID 10737
IUPAC Name [4-(dimethylamino)phenyl]-phenylmethanone
SMILES CN(C)C1=CC=C(C=C1)C(=O)C2=CC=CC=C2

Specifications

Melting Point 88°C to 90°C
Quantity 5 g
Merck Index 14,3233
Solubility Information Insoluble in water.
Formula Weight 225.29
Percent Purity 98%
Chemical Name or Material 4-Dimethylaminobenzophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-478-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only