accessibility menu, dialog, popup

UserName

CAS RN 530-44-9

IUPAC Name: 4-benzoyl-N,N-dimethylaniline
Synonyms: 4-benzoyl-N,N-dimethylaniline
Molecular Weight (g/mol): 225.29
Molecular Formula: C15H15NO
InChi Key: BEUGBYXJXMVRFO-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)C(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)