missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylamino)phenyldiphenylphosphine 95.0+%, TCI America™

Catalog No. p-7127321
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24781G 1 g
D24785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D24781G Supplier TCI America Supplier No. D24781G
Only null left

Chemical Identifiers

CAS 739-58-2
Molecular Formula C20H20NP
Molecular Weight (g/mol) 305.36
MDL Number MFCD00192068
InChI Key GOEGBJDTWXTPHP-UHFFFAOYSA-N
Synonym 4-dimethylamino phenyldiphenylphosphine, 4-diphenylphosphino-n,n-dimethylaniline, 4-diphenylphosphanyl-n,n-dimethylaniline, 4-dimethylamino triphenylphosphine, benzenamine, 4-diphenylphosphino-n,n-dimethyl, 4-dimethylaminophenyl diphenylphosphine, 4-diphenylphosphino-n,n-dimethyl-benzenamine, acmc-209ot0, ksc495a7n, p-dimethylaminophenyl-diphenyl-phosphine
PubChem CID 3333592
IUPAC Name 4-(diphenylphosphanyl)-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 152°C
Color White
Quantity 1 g
Formula Weight 305.36
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Dimethylamino)phenyldiphenylphosphine