accessibility menu, dialog, popup

UserName

CAS RN 739-58-2

H3CNCH3P

CAS RN 739-58-2

IUPAC Name: 4-(diphenylphosphanyl)-N,N-dimethylaniline
Synonyms: 4-(diphenylphosphanyl)-N,N-dimethylaniline
Molecular Weight (g/mol): 305.36
Molecular Formula: C20H20NP
InChi Key: GOEGBJDTWXTPHP-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results