missing translation for 'onlineSavingsMsg'
Learn More

4-(Difluoromethoxy)aniline 98.0+%, TCI America™

Catalog No. D34575G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D34575G Supplier TCI America Supplier No. D34575G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 22236-10-8
Molecular Formula C7H7F2NO
Molecular Weight (g/mol) 159.136
MDL Number MFCD00085005
InChI Key NDEZTSHWEPQVBX-UHFFFAOYSA-N
Synonym 4-difluoromethoxy aniline, 4-difluoromethoxy-phenylamine, p-difluoromethoxyaniline, 4-amino-alpha,alpha-difluoroanisole, 4-difluoromethoxy benzenamine, benzenamine, 4-difluoromethoxy, 4-difluoromethoxy phenylamine, 4-difluormethoxy anilin, pubchem8467
PubChem CID 737363
IUPAC Name 4-(difluoromethoxy)aniline
SMILES C1=CC(=CC=C1N)OC(F)F

Specifications

Color Yellow
Boiling Point 90°C
Quantity 5 g
Formula Weight 159.14
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-(Difluoromethoxy)aniline