accessibility menu, dialog, popup

UserName

CAS RN 22236-10-8

IUPAC Name: 4-(difluoromethoxy)aniline
Synonyms: 4-(difluoromethoxy)aniline
Molecular Weight (g/mol): 159.14
Molecular Formula: C7H7F2NO
InChi Key: NDEZTSHWEPQVBX-UHFFFAOYSA-N
SMILES: NC1=CC=C(OC(F)F)C=C1
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)