missing translation for 'onlineSavingsMsg'
Learn More

4-[Difluoro(3,4,5-trifluorophenoxy)methyl]-3,5-difluoro-4'-propylbiphenyl 98.0+%, TCI America™

Catalog No. D51305G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D51305G 5 g
D51301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D51305G Supplier TCI America Supplier No. D51305G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 303186-20-1
Molecular Formula C22H15F7O
Molecular Weight (g/mol) 428.35
MDL Number MFCD18451990
InChI Key YJUHXJQZJUQYJJ-UHFFFAOYSA-N
Synonym 4-difluoro 3,4,5-trifluorophenoxy methyl-3,5-difluoro-4'-propyl-1,1'-biphenyl, 4-difluoro 3,4,5-trifluorophenoxy methyl-3,5-difluoro-4'-propylbiphenyl, 4-3,4,5-trifluorophenoxydifluoromethyl-3,5-difluoro-4'-propylbiphenyl, 2-difluoro-3,4,5-trifluorophenoxy methyl-1,3-difluoro-5-4-pro pylphenyl benzene, 4-difluoro 3,4,5-trifluorophenoxy methyl-3,5-difluoro-4-propyl-1,1-biphenyl, 4-difluoro 3,4,5-trifluorophenoxy methyl-3,5-difluoro-4'-n-propylbiphenyl
PubChem CID 20654145
IUPAC Name 2-[difluoro-(3,4,5-trifluorophenoxy)methyl]-1,3-difluoro-5-(4-propylphenyl)benzene
SMILES CCCC1=CC=C(C=C1)C2=CC(=C(C(=C2)F)C(OC3=CC(=C(C(=C3)F)F)F)(F)F)F

Specifications

Melting Point 49°C
Color White
Quantity 5 g
Formula Weight 428.35
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-[Difluoro(3,4,5-trifluorophenoxy)methyl]-3,5-difluoro-4′-propylbiphenyl
EINECSNumber (4)-1935
TSCA No