missing translation for 'onlineSavingsMsg'
Learn More

4-Diethylaminobenzaldehyde 99.0+%, TCI America™

Catalog No. p-7126228
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
D046325G 25 g
D0463100G 100 g
D0463500G 500 g
3 options

Catalog No. D046325G

Supplier: TCI America D046325G

Only null left

Chemical Identifiers

CAS 120-21-8
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
MDL Number MFCD00003382
InChI Key MNFZZNNFORDXSV-UHFFFAOYSA-N
Synonym 4-diethylamino benzaldehyde, benzaldehyde, 4-diethylamino, p-diethylamino benzaldehyde, p-formyl-n,n-diethylaniline, 4-diethylamino-benzaldehyde, benzaldehyde, p-diethylamino, unii-s640xwl10h, p-n,n-diethylaminobenzaldehyde, p-diethylaminobenzaldehyde, 4-n,n-diethylamino benzaldehyde
PubChem CID 67114
ChEBI CHEBI:86194
IUPAC Name 4-(diethylamino)benzaldehyde
SMILES CCN(CC)C1=CC=C(C=C1)C=O

Specifications

Melting Point 42°C
Boiling Point 174°C
Quantity 25 g
Formula Weight 177.25
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Diethylaminobenzaldehyde
EINECSNumber (3)-1170
RTECSNumber CU5612850
TSCA Yes
Recommended Storage Refrigerator