accessibility menu, dialog, popup

UserName

CH3NCH3O

CAS RN 120-21-8

IUPAC Name: 4-(diethylamino)benzaldehyde
Synonyms: 4-(diethylamino)benzaldehyde
Molecular Weight (g/mol): 177.25
Molecular Formula: C11H15NO
InChi Key: MNFZZNNFORDXSV-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C(C=O)C=C1
Boiling Point 
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (3)