missing translation for 'onlineSavingsMsg'
Learn More

4-Diethylaminobenzaldehyde, 97%

Catalog No. p-7081730
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
Catalog No. Quantity
AAA1182522 100 g
AAA1182536 500 g
2 options

Catalog No. AAA1182522

Supplier: Thermo Scientific Chemicals A1182522

Only null left

CAS: 120-21-8 | C11H15NO | 177.247 g/mol

4-Diethylaminobenzaldehyde is used in the preparation of 5-(4-diethylamino-benzylidene)-selenazolidine-2,4-dione by using aqueous methylamine as a reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Diethylaminobenzaldehyde is used in the preparation of 5-(4-diethylamino-benzylidene)-selenazolidine-2,4-dione by using aqueous methylamine as a reagent.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 120-21-8
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
MDL Number MFCD00003382
InChI Key MNFZZNNFORDXSV-UHFFFAOYSA-N
Synonym 4-diethylamino benzaldehyde, benzaldehyde, 4-diethylamino, p-diethylamino benzaldehyde, p-formyl-n,n-diethylaniline, 4-diethylamino-benzaldehyde, benzaldehyde, p-diethylamino, unii-s640xwl10h, p-n,n-diethylaminobenzaldehyde, p-diethylaminobenzaldehyde, 4-n,n-diethylamino benzaldehyde
PubChem CID 67114
ChEBI CHEBI:86194
IUPAC Name 4-(diethylamino)benzaldehyde
SMILES CCN(CC)C1=CC=C(C=C1)C=O

Specifications

Melting Point 39°C to 41°C
Boiling Point 173°C to 175°C (7 mmHg)
Flash Point 165°C (329°F)
Quantity 100 g
Beilstein 511102
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 177.25
Percent Purity 97%
Chemical Name or Material 4-Diethylaminobenzaldehyde
Hazard Category H311-H315-H319-H335
Hazard Statement GHS H Statement
H311-H315-H319-H335
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 204-377-4
RTECSNumber CU5612850
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only