missing translation for 'onlineSavingsMsg'
Learn More

4-(Diethylamino)benzaldehyde, 98+%

Catalog No. AC114001000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC114001000 100 g Glass bottle
AC114005000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC114001000 Supplier Thermo Scientific Chemicals Supplier No. 114001000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 120-21-8 | C11H15NO | 177.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 120-21-8
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.25
MDL Number MFCD00003382
InChI Key MNFZZNNFORDXSV-UHFFFAOYSA-N
Synonym 4-diethylamino benzaldehyde, benzaldehyde, 4-diethylamino, p-diethylamino benzaldehyde, p-formyl-n,n-diethylaniline, 4-diethylamino-benzaldehyde, benzaldehyde, p-diethylamino, unii-s640xwl10h, p-n,n-diethylaminobenzaldehyde, p-diethylaminobenzaldehyde, 4-n,n-diethylamino benzaldehyde
PubChem CID 67114
ChEBI CHEBI:86194
IUPAC Name 4-(diethylamino)benzaldehyde
SMILES CCN(CC)C1=CC=C(C=C1)C=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 38.0°C to 41.0°C
Color Black to Brown
Boiling Point 174.0°C (7.0 mmHg)
Flash Point 165°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula (C2H5)2NC6H4CHO
Quantity 100 g
Beilstein 14,II,25
Solubility Information Solubility in water: 2.5g/L (20°C). Other solubilities: soluble in methanol
Formula Weight 177.25
Percent Purity 98+%
Physical Form Coarse Crystals, Chunks or Crystalline Solid
Chemical Name or Material 4-(Diethylamino)benzaldehyde, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
EINECSNumber 204-377-4
RTECSNumber CU5612850
TSCA TSCA

RUO – Research Use Only