missing translation for 'onlineSavingsMsg'
Learn More

4-(Decyloxy)benzoic Acid 98.0+%, TCI America™

Catalog No. D41765G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41765G 5 g
D417625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41765G Supplier TCI America Supplier No. D41765G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5519-23-3
Molecular Formula C17H26O3
Molecular Weight (g/mol) 278.392
MDL Number MFCD00020360
InChI Key NZNICZRIRMGOFG-UHFFFAOYSA-N
Synonym 4-decyloxy benzoic acid, p-decyloxybenzoic acid, 4-n-decyloxybenzoic acid, 4-decyloxybenzoic acid, benzoic acid,4-decyloxy, benzoic acid, 4-decyloxy, pubchem2671, p-n-decyloxybenzoic acid, 4-decyloxy benzoicacid, para-decyloxy benzoic acid
PubChem CID 138527
IUPAC Name 4-decoxybenzoic acid
SMILES CCCCCCCCCCOC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 96°C
Color White
Quantity 5 g
Formula Weight 278.39
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Decyloxy)benzoic Acid