Learn More
Chemical Identifiers
| CAS | 4395-98-6 |
|---|---|
| Molecular Formula | C6H10N2 |
| Molecular Weight (g/mol) | 110.16 |
| InChI Key | FSDNTQSJGHSJBG-UHFFFAOYSA-N |
| Synonym | 4-cyanopiperidine, 4-piperidinecarbonitrile, 4-cyanopiperdine, 4-cyanopyperidine, 4-cyano piperdine, 4-cyano piperidine, 4-cyano-piperidine, acmc-20aipl, pubchem9349 |
| PubChem CID | 138223 |
| IUPAC Name | piperidine-4-carbonitrile |
| SMILES | C1CNCCC1C#N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC441820050
|
N/A | 5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC441820250
|
N/A | 25g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 4395-98-6 | |
| 110.16 | |
| 4-cyanopiperidine, 4-piperidinecarbonitrile, 4-cyanopiperdine, 4-cyanopyperidine, 4-cyano piperdine, 4-cyano piperidine, 4-cyano-piperidine, acmc-20aipl, pubchem9349 | |
| piperidine-4-carbonitrile |
| C6H10N2 | |
| FSDNTQSJGHSJBG-UHFFFAOYSA-N | |
| 138223 | |
| C1CNCCC1C#N |
Specifications
| 4395-98-6 | |
| 100.0 | |
| Authentic | |
| C6H10N2 | |
| FSDNTQSJGHSJBG-UHFFFAOYSA-N | |
| piperidine-4-carbonitrile | |
| 138223 | |
| 97% |
| 96.0 | |
| 100.0°C to 105.0°C (12.0 mmHg) | |
| 96% min. (GC) | |
| 4-cyanopiperidine, 4-piperidinecarbonitrile, 4-cyanopiperdine, 4-cyanopyperidine, 4-cyano piperdine, 4-cyano piperidine, 4-cyano-piperidine, acmc-20aipl, pubchem9349 | |
| C1CNCCC1C#N | |
| 110.16 | |
| 110.16 | |
| 4-Cyanopiperidine |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Danger
RUO – Research Use Only