accessibility menu, dialog, popup

UserName

NNH

CAS RN 4395-98-6

IUPAC Name: piperidine-4-carbonitrile
Synonyms: piperidine-4-carbonitrile
Molecular Weight (g/mol): 110.16
Molecular Formula: C6H10N2
InChi Key: FSDNTQSJGHSJBG-UHFFFAOYSA-N
SMILES: N#CC1CCNCC1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)