missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanopiperidine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 441820250

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC441820250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

4395-98-6
110.16
4-cyanopiperidine, 4-piperidinecarbonitrile, 4-cyanopiperdine, 4-cyanopyperidine, 4-cyano piperdine, 4-cyano piperidine, 4-cyano-piperidine, acmc-20aipl, pubchem9349
piperidine-4-carbonitrile
C6H10N2
FSDNTQSJGHSJBG-UHFFFAOYSA-N
138223
C1CNCCC1C#N

Specifications

4-Cyanopiperidine
96.0
100.0°C to 105.0°C (12.0 mmHg)
96% min. (GC)
C6H10N2
4-cyanopiperidine, 4-piperidinecarbonitrile, 4-cyanopiperdine, 4-cyanopyperidine, 4-cyano piperdine, 4-cyano piperidine, 4-cyano-piperidine, acmc-20aipl, pubchem9349
C1CNCCC1C#N
110.16
110.16
4395-98-6
100.0
Authentic
Glass bottle
25g
FSDNTQSJGHSJBG-UHFFFAOYSA-N
piperidine-4-carbonitrile
138223
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Danger

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...