missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanophenylacetonitrile, 97%

Catalog No. p-7035887
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5411203 1 g
AAH5411206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5411203 Supplier Thermo Scientific Chemicals Supplier No. H5411203
Only null left

CAS: 876-31-3 | C9H6N2 | 142.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 876-31-3
Molecular Formula C9H6N2
Molecular Weight (g/mol) 142.16
MDL Number MFCD00060305
InChI Key QILKKAFYAFEWGU-UHFFFAOYSA-N
Synonym 4-cyanomethyl benzonitrile, 4-cyanophenylacetonitrile, benzeneacetonitrile, 4-cyano, p-tolunitrile, .alpha.-cyano, p-cyanobenzylcyanide, 4-cyanobenzeneacetonitrile, .alpha.-cyano-p-tolunitrile, 4-cyanbenzylcyanide, 4-cyanobenzylcyanide, acmc-209qoz
PubChem CID 136686
IUPAC Name 4-(cyanomethyl)benzonitrile
SMILES N#CCC1=CC=C(C=C1)C#N

Specifications

Density 1.056
Boiling Point 180°C to 181°C
Flash Point 71°C (160°F)
Refractive Index 1.499
Quantity 1 g
Formula Weight 142.16
Percent Purity 97%
Chemical Name or Material 4-Cyanophenylacetonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only