missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobenzyl Chloride 97.0+%, TCI America™

Catalog No. p-7125222
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C19855G 5 g
C198525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C19855G Supplier TCI America Supplier No. C19855G
Only null left

Chemical Identifiers

CAS 874-86-2
Molecular Formula C8H6ClN
Molecular Weight (g/mol) 151.593
InChI Key LOQLDQJTSMKBJU-UHFFFAOYSA-N
Synonym 4-chloromethyl benzonitrile, 4-cyanobenzyl chloride, 4-chloromethyl-benzonitrile, benzonitrile, 4-chloromethyl, ccris 5103, p-cyanobenzyl chloride, 4-chloromethyl benzenecarbonitrile, 4-cyanobenzylchloride, pubchem7377, alp.-chloro-p-tolunitrile
PubChem CID 70127
IUPAC Name 4-(chloromethyl)benzonitrile
SMILES C1=CC(=CC=C1CCl)C#N

Specifications

Melting Point 80°C
Color White-Yellow
Boiling Point 263°C
Quantity 5 g
UN Number 2923
Formula Weight 151.59
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyanobenzyl Chloride
EINECSNumber (3)-1810
TSCA No