missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobenzoic acid, 99%

Catalog No. AC110730050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC110730050 5 g Glass bottle
AC110730250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC110730050 Supplier Thermo Scientific Chemicals Supplier No. 110730050

CAS: 619-65-8 | C8H5NO2 | 147.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 619-65-8
Molecular Formula C8H5NO2
Molecular Weight (g/mol) 147.13
MDL Number MFCD00002528
InChI Key ADCUEPOHPCPMCE-UHFFFAOYSA-N
Synonym p-cyanobenzoic acid, benzoic acid, 4-cyano, p-carboxybenzonitrile, 4-carboxybenzonitrile, benzoic acid, p-cyano, 4-cyano-benzoic acid, 4-cyano benzoic acid, benzoicacid, 4-cyano, 4-cyanobenzoicacid, para-cyanobenzoic acid
PubChem CID 12087
IUPAC Name 4-cyanobenzoic acid
SMILES C1=CC(=CC=C1C#N)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 217.0°C to 222.0°C
Color Beige-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula NCC6H4CO2H
Quantity 5 g
Beilstein 09, 845
Solubility Information Solubility in water: 1.2g/L (20°C). Other solubilities: soluble in methanol
Formula Weight 147.13
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyanobenzoic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove
EINECSNumber 210-606-9
TSCA TSCA

RUO – Research Use Only