missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobenzoic Acid 98.0+%, TCI America™

Catalog No. C04455G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C04455G 5 g
C044525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C04455G Supplier TCI America Supplier No. C04455G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 619-65-8
Molecular Formula C8H5NO2
Molecular Weight (g/mol) 147.133
MDL Number MFCD00002528
InChI Key ADCUEPOHPCPMCE-UHFFFAOYSA-N
Synonym p-cyanobenzoic acid, benzoic acid, 4-cyano, p-carboxybenzonitrile, 4-carboxybenzonitrile, benzoic acid, p-cyano, 4-cyano-benzoic acid, 4-cyano benzoic acid, benzoicacid, 4-cyano, 4-cyanobenzoicacid, para-cyanobenzoic acid
PubChem CID 12087
IUPAC Name 4-cyanobenzoic acid
SMILES C1=CC(=CC=C1C#N)C(=O)O

Specifications

Melting Point 224°C
Color White
Quantity 5 g
Formula Weight 147.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyanobenzoic Acid
EINECSNumber (9)-0528
TSCA Yes