missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Cyano-4'-n-octylbiphenyl 98.0+%, TCI America™
Supplier: TCI America C315625G
Specifications
| 4-Cyano-4′-n-octylbiphenyl | |
| White | |
| C21H25N | |
| 25 g | |
| CSQPODPWWMOTIY-UHFFFAOYSA-N | |
| 4'-octyl-[1,1'-biphenyl]-4-carbonitrile | |
| 104289 | |
| ≥98.0% (GC) |
| 52709-84-9 | |
| 218°C | |
| MFCD00075146 | |
| 4-cyano-4'-octylbiphenyl, 4'-octyl-1,1'-biphenyl-4-carbonitrile, 4-4-octylphenyl benzonitrile, 1,1'-biphenyl-4-carbonitrile, 4'-octyl, 4-octylcyanodiphenyl, k 24 liquid crystal, cb 8, 4'-octyl-4-biphenylcarbonitrile, 4'-octyl 1,1'-biphenyl-4-carbonitrile, 4-octyl-4'-cyanobiphenyl | |
| CCCCCCCCC1=CC=C(C=C1)C1=CC=C(C=C1)C#N | |
| 291.44 | |
| 291.44 | |
| Crystalline Powder |
Chemical Identifiers
| 52709-84-9 | |
| 291.44 | |
| CSQPODPWWMOTIY-UHFFFAOYSA-N | |
| 104289 | |
| CCCCCCCCC1=CC=C(C=C1)C1=CC=C(C=C1)C#N |
| C21H25N | |
| MFCD00075146 | |
| 4-cyano-4'-octylbiphenyl, 4'-octyl-1,1'-biphenyl-4-carbonitrile, 4-4-octylphenyl benzonitrile, 1,1'-biphenyl-4-carbonitrile, 4'-octyl, 4-octylcyanodiphenyl, k 24 liquid crystal, cb 8, 4'-octyl-4-biphenylcarbonitrile, 4'-octyl 1,1'-biphenyl-4-carbonitrile, 4-octyl-4'-cyanobiphenyl | |
| 4'-octyl-[1,1'-biphenyl]-4-carbonitrile |
Safety and Handling
RTECSNumber : DV1764400
TSCA : Yes