missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-4'-heptyloxybiphenyl 98.0+%, TCI America™

Catalog No. C16061G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C16061G Supplier TCI America Supplier No. C16061G
Only null left

Chemical Identifiers

CAS 52364-72-4
Molecular Formula C20H23NO
Molecular Weight (g/mol) 293.41
MDL Number MFCD00134146
InChI Key JPBFKTCKZLMJED-UHFFFAOYSA-N
Synonym 4'-heptyloxy-4-cyanobiphenyl, 4-cyano-4'-heptyloxybiphenyl, 4'-heptyloxy-1,1'-biphenyl-4-carbonitrile, 4'-heptyloxy biphenyl-4-carbonitrile, 7cob, 1,1'-biphenyl-4-carbonitrile, 4'-heptyloxy, 4-heptyloxycyanodiphenyl, 4-cyano-4'-heptoxybiphenyl, 4-4-heptoxyphenyl benzonitrile, 4-4-heptyloxyphenyl benzonitrile
PubChem CID 104172
IUPAC Name 4-(4-heptoxyphenyl)benzonitrile
SMILES CCCCCCCOC1=CC=C(C=C1)C2=CC=C(C=C2)C#N

Specifications

Melting Point 54°C
Color White-Yellow
Quantity 1 g
Formula Weight 293.41
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyano-4′-heptyloxybiphenyl
RTECSNumber DV1764100
TSCA Yes