accessibility menu, dialog, popup

UserName

H3CON

CAS RN 52364-72-4

IUPAC Name: 4'-(heptyloxy)-[1,1'-biphenyl]-4-carbonitrile
Synonyms: 4'-(heptyloxy)-[1,1'-biphenyl]-4-carbonitrile
Molecular Weight (g/mol): 293.41
Molecular Formula: C20H23NO
InChi Key: JPBFKTCKZLMJED-UHFFFAOYSA-N
SMILES: CCCCCCCOC1=CC=C(C=C1)C1=CC=C(C=C1)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results