missing translation for 'onlineSavingsMsg'
Learn More

4'-Cyano-4-dodecyloxybiphenyl 98.0+%, TCI America™

Catalog No. C32701G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C32701G 1 g
C32705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C32701G Supplier TCI America Supplier No. C32701G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57125-50-5
Molecular Formula C25H33NO
Molecular Weight (g/mol) 363.545
MDL Number MFCD01941019
InChI Key WYSZDBDWXPAHDS-UHFFFAOYSA-N
PubChem CID 92674
IUPAC Name 4-(4-dodecoxyphenyl)benzonitrile
SMILES CCCCCCCCCCCCOC1=CC=C(C=C1)C2=CC=C(C=C2)C#N

Specifications

Color White
Quantity 1 g
Synonym 4′C-Dodecyloxy-4-biphenylcarbonitrile, 4′C-(Dodecyloxy)-[1,1′C-biphenyl]-4-carbonitrile, 12OCB
Formula Weight 363.55
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Cyano-4-dodecyloxybiphenyl