accessibility menu, dialog, popup

UserName

CAS RN 57125-50-5

IUPAC Name: 4'-(dodecyloxy)-[1,1'-biphenyl]-4-carbonitrile
Synonyms: 4'-(dodecyloxy)-[1,1'-biphenyl]-4-carbonitrile
Molecular Weight (g/mol): 363.55
Molecular Formula: C25H33NO
InChi Key: WYSZDBDWXPAHDS-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCOC1=CC=C(C=C1)C1=CC=C(C=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results