missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Cyano-2-fluorobenzoic Acid 97.0+%, TCI America™
Supplier: TCI America C20515G
Specifications
| 4-Cyano-2-fluorobenzoic Acid | |
| 216°C | |
| C8H4FNO2 | |
| 5 g | |
| 2-fluoro-4-cyanobenzoic acid, benzoic acid, 4-cyano-2-fluoro, 4-cyano-2-fluoro-benzoic acid, 4-cyano-2-fluorobenzoicacid, pubchem4603, acmc-1bzbk, intermediates-zcf02232, 2-fluoro4-cyanobenzoic acid, ksc489o1d, 4-carboxy-3-fluorobenzonitrile | |
| OC(=O)C1=C(F)C=C(C=C1)C#N | |
| 165.12 | |
| 165.12 | |
| Crystalline Powder |
| 164149-28-4 | |
| White | |
| MFCD03094454 | |
| 3439 | |
| KEJMSTJTAWACNI-UHFFFAOYSA-N | |
| 4-cyano-2-fluorobenzoic acid | |
| 590659 | |
| ≥97.0% (GC,T) |
Chemical Identifiers
| 164149-28-4 | |
| 165.12 | |
| KEJMSTJTAWACNI-UHFFFAOYSA-N | |
| 590659 | |
| OC(=O)C1=C(F)C=C(C=C1)C#N |
| C8H4FNO2 | |
| MFCD03094454 | |
| 2-fluoro-4-cyanobenzoic acid, benzoic acid, 4-cyano-2-fluoro, 4-cyano-2-fluoro-benzoic acid, 4-cyano-2-fluorobenzoicacid, pubchem4603, acmc-1bzbk, intermediates-zcf02232, 2-fluoro4-cyanobenzoic acid, ksc489o1d, 4-carboxy-3-fluorobenzonitrile | |
| 4-cyano-2-fluorobenzoic acid |
Safety and Handling
TSCA : No