Learn More
(4-Chlorophenylthio)acetic acid, 98%
CAS: 3405-88-7 | C8H7ClO2S | 202.652 g/mol
Supplier: Thermo Scientific Chemicals L0858318
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| (4-Chlorophenylthio)acetic acid | |
| 104°C to 107°C | |
| MFCD00021762 | |
| 1871569 | |
| YPKLXLYGMAWXDO-UHFFFAOYSA-N | |
| 2-(4-chlorophenyl)sulfanylacetic acid | |
| 55471 | |
| 98% |
| 3405-88-7 | |
| C8H7ClO2S | |
| 50 g | |
| 4-chlorophenylthio acetic acid, 2-4-chlorophenyl thio acetic acid, 4-chlorophenyl thio acetic acid, p-chlorophenylmercaptoacetic acid, 2-4-chlorophenyl sulfanyl acetic acid, 2-p-chlorophenylthio acetic acid, p-chlorophenylthioacetic acid, 4-chlorophenyl sulfanyl acetic acid, acetic acid, p-chlorophenyl thio, acetic acid, 4-chlorophenyl thio | |
| C1=CC(=CC=C1SCC(=O)O)Cl | |
| 202.652 | |
| 202.66 |
Chemical Identifiers
| 3405-88-7 | |
| 202.652 | |
| YPKLXLYGMAWXDO-UHFFFAOYSA-N | |
| 55471 | |
| C1=CC(=CC=C1SCC(=O)O)Cl |
| C8H7ClO2S | |
| MFCD00021762 | |
| 4-chlorophenylthio acetic acid, 2-4-chlorophenyl thio acetic acid, 4-chlorophenyl thio acetic acid, p-chlorophenylmercaptoacetic acid, 2-4-chlorophenyl sulfanyl acetic acid, 2-p-chlorophenylthio acetic acid, p-chlorophenylthioacetic acid, 4-chlorophenyl sulfanyl acetic acid, acetic acid, p-chlorophenyl thio, acetic acid, 4-chlorophenyl thio | |
| 2-(4-chlorophenyl)sulfanylacetic acid |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
RTECSNumber : AG1075000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only