missing translation for 'onlineSavingsMsg'
Learn More

(4-Chlorophenylthio)acetic acid, 98%

Catalog No. p-7044877
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0858309 10 g
AAL0858318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0858309 Supplier Thermo Scientific Chemicals Supplier No. L0858309

CAS: 3405-88-7 | C8H7ClO2S | 202.652 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3405-88-7
Molecular Formula C8H7ClO2S
Molecular Weight (g/mol) 202.652
MDL Number MFCD00021762
InChI Key YPKLXLYGMAWXDO-UHFFFAOYSA-N
Synonym 4-chlorophenylthio acetic acid, 2-4-chlorophenyl thio acetic acid, 4-chlorophenyl thio acetic acid, p-chlorophenylmercaptoacetic acid, 2-4-chlorophenyl sulfanyl acetic acid, 2-p-chlorophenylthio acetic acid, p-chlorophenylthioacetic acid, 4-chlorophenyl sulfanyl acetic acid, acetic acid, p-chlorophenyl thio, acetic acid, 4-chlorophenyl thio
PubChem CID 55471
IUPAC Name 2-(4-chlorophenyl)sulfanylacetic acid
SMILES C1=CC(=CC=C1SCC(=O)O)Cl

Specifications

Melting Point 104°C to 107°C
Quantity 10 g
Beilstein 1871569
Formula Weight 202.66
Percent Purity 98%
Chemical Name or Material (4-Chlorophenylthio)acetic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
RTECSNumber AG1075000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only