missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorophenylhydrazine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America C0257250G
Specifications
| 4-Chlorophenylhydrazine Hydrochloride | |
| C6H8Cl2N2 | |
| 250 g | |
| YQVZREHUWCCHHX-UHFFFAOYSA-N | |
| (4-chlorophenyl)hydrazine;hydrochloride | |
| 71600 | |
| ≥98.0% (HPLC,T) |
| 1073-70-7 | |
| MFCD00012943 | |
| 4-chlorophenylhydrazine hydrochloride, 4-chlorophenyl hydrazine hydrochloride, p-chlorophenylhydrazine hydrochloride, p-chlorophenylhydrazine hcl, 4-chlorophenylhydrazine hcl, 1-4-chlorophenyl hydrazine hydrochloride, 4-chloro-phenyl-hydrazine hydrochloride, chlorophenylhydrazine 4-hcl, hydrazine, 4-chlorophenyl-, monohydrochloride, 4-chlorophenylhydrazine, chloride | |
| C1=CC(=CC=C1NN)Cl.Cl | |
| 179.044 | |
| 179.04 | |
| Crystalline Powder |
Chemical Identifiers
| 1073-70-7 | |
| 179.044 | |
| YQVZREHUWCCHHX-UHFFFAOYSA-N | |
| 71600 | |
| C1=CC(=CC=C1NN)Cl.Cl |
| C6H8Cl2N2 | |
| MFCD00012943 | |
| 4-chlorophenylhydrazine hydrochloride, 4-chlorophenyl hydrazine hydrochloride, p-chlorophenylhydrazine hydrochloride, p-chlorophenylhydrazine hcl, 4-chlorophenylhydrazine hcl, 1-4-chlorophenyl hydrazine hydrochloride, 4-chloro-phenyl-hydrazine hydrochloride, chlorophenylhydrazine 4-hcl, hydrazine, 4-chlorophenyl-, monohydrochloride, 4-chlorophenylhydrazine, chloride | |
| (4-chlorophenyl)hydrazine;hydrochloride |
Safety and Handling
TSCA : No