missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorophenylhydrazine hydrochloride, 97%
Description
4-Chlorophenylhydrazine hydrochloride is used as a pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Chlorophenylhydrazine hydrochloride is used as a pharmaceutical intermediates.
Solubility
Soluble in hot water, methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
4-Chlorophenylhydrazine hydrochloride is used as a pharmaceutical intermediates.
Solubility
Soluble in hot water, methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | ∼216°C (decomposition) |
| Odor | Odorless |
| Quantity | 25 g |
| Beilstein | 3596254 |
| Solubility Information | Soluble in hot water,methanol. |
| Formula Weight | 179.05 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-Chlorophenylhydrazine hydrochloride |
Safety and Handling
| missing translation for 'healthHazard1' | H302+H312+H332 |
| missing translation for 'healthHazard2' | GHS H Statement H301-H350-H315-H319-H335 Toxic if swallowed. May cause cancer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| missing translation for 'healthHazard3' | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c |
| EINECSNumber | 214-030-9 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only