missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(4-Chlorophenyl)thiourea 98.0+%, TCI America™
Supplier: TCI America C296025G
Specifications
| (4-Chlorophenyl)thiourea | |
| 171°C | |
| C7H7ClN2S | |
| 25 g | |
| 1-4-chlorophenyl thiourea, 4-chlorophenyl thiourea, n-4-chlorophenyl thiourea, thiourea, 4-chlorophenyl, 1-p-chlorophenyl thiourea, n-p-chlorophenyl thiourea, 1-p-chlorophenyl-2-thiourea, 4-chlorophenyl-2-thiourea, urea, 1-p-chlorophenyl-2-thio | |
| NC(=S)NC1=CC=C(Cl)C=C1 | |
| 186.66 | |
| 186.66 | |
| Crystalline Powder |
| 3696-23-9 | |
| White-Yellow | |
| MFCD00022168 | |
| 2811 | |
| XVEFWRUIYOXUGG-UHFFFAOYSA-N | |
| (4-chlorophenyl)thiourea | |
| 735843 | |
| ≥98.0% (HPLC,N) |
Chemical Identifiers
| 3696-23-9 | |
| 186.66 | |
| XVEFWRUIYOXUGG-UHFFFAOYSA-N | |
| 735843 | |
| NC(=S)NC1=CC=C(Cl)C=C1 |
| C7H7ClN2S | |
| MFCD00022168 | |
| 1-4-chlorophenyl thiourea, 4-chlorophenyl thiourea, n-4-chlorophenyl thiourea, thiourea, 4-chlorophenyl, 1-p-chlorophenyl thiourea, n-p-chlorophenyl thiourea, 1-p-chlorophenyl-2-thiourea, 4-chlorophenyl-2-thiourea, urea, 1-p-chlorophenyl-2-thio | |
| (4-chlorophenyl)thiourea |
Safety and Handling
RTECSNumber : YS7175000
TSCA : No