missing translation for 'onlineSavingsMsg'
Learn More

(4-Chlorophenyl)thiourea 98.0+%, TCI America™

Catalog No. p-7125746
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29605G 5 g
C296025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29605G Supplier TCI America Supplier No. C29605G
Only null left

Chemical Identifiers

CAS 3696-23-9
Molecular Formula C7H7ClN2S
Molecular Weight (g/mol) 186.66
MDL Number MFCD00022168
InChI Key XVEFWRUIYOXUGG-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl thiourea, 4-chlorophenyl thiourea, n-4-chlorophenyl thiourea, thiourea, 4-chlorophenyl, 1-p-chlorophenyl thiourea, n-p-chlorophenyl thiourea, 1-p-chlorophenyl-2-thiourea, 4-chlorophenyl-2-thiourea, urea, 1-p-chlorophenyl-2-thio
PubChem CID 735843
IUPAC Name (4-chlorophenyl)thiourea
SMILES NC(=S)NC1=CC=C(Cl)C=C1

Specifications

Melting Point 171°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 186.66
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (4-Chlorophenyl)thiourea
RTECSNumber YS7175000
TSCA No