missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenyl Phosphorodichloridate 97.0+%, TCI America™

Catalog No. p-7124718
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
C09775G 5 g
C097725G 25 g
2 options

Catalog No. C09775G

Supplier: TCI America C09775G

Only null left

Chemical Identifiers

CAS 772-79-2
Molecular Formula C6H4Cl3O2P
Molecular Weight (g/mol) 245.42
MDL Number MFCD00009705
InChI Key CCZMQYGSXWZFKI-UHFFFAOYSA-N
Synonym 4-chlorophenyl phosphorodichloridate, p-chlorophenyl dichlorophosphate, 4-chlorophenyl dichlorophosphate, 4-chlorophenylphosphorodichloridate, phosphorodichloridic acid, 4-chlorophenyl ester, 4-chlorophenylphosphoryl dichloride, acmc-1bjgo, cczmqygsxwzfki-uhfffaoysa, 4-chlorophenyl dichlorophosphinate, 4-chlorophenyl dichloridophosphate #
PubChem CID 69879
IUPAC Name 1-chloro-4-dichlorophosphoryloxybenzene
SMILES C1=CC(=CC=C1OP(=O)(Cl)Cl)Cl

Specifications

Color Yellow
Boiling Point 265°C
Quantity 5 g
UN Number 3265
Formula Weight 245.42
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material 4-Chlorophenyl Phosphorodichloridate [Phosphorylating Agent]