accessibility menu, dialog, popup

UserName

ClOPClClO

CAS RN 772-79-2

IUPAC Name: 4-chlorophenyl chlorophosphonochloridate
Synonyms: 4-chlorophenyl chlorophosphonochloridate
Molecular Weight (g/mol): 245.42
Molecular Formula: C6H4Cl3O2P
InChi Key: CCZMQYGSXWZFKI-UHFFFAOYSA-N
SMILES: ClC1=CC=C(OP(Cl)(Cl)=O)C=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)