missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenyl phosphorodichloridate, 98+%

Catalog No. AAA1550714
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1550714 25 g
AAA1550722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1550714 Supplier Thermo Scientific Chemicals Supplier No. A1550714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 772-79-2 | C6H4Cl3O2P | 245.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-Chlorophenyl phosphorodichloridate is used as phosphorylation agent. Reactant for preparation of antimicrobial agents, antioxidant agents, antitumor agents, nucleotide anti-viral prodrug.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store under inert gas. Store away from moisture, oxidizing agents, alcohols, bases.

Chemical Identifiers

CAS 772-79-2
Molecular Formula C6H4Cl3O2P
Molecular Weight (g/mol) 245.42
MDL Number MFCD00009705
InChI Key CCZMQYGSXWZFKI-UHFFFAOYSA-N
Synonym 4-chlorophenyl phosphorodichloridate, p-chlorophenyl dichlorophosphate, 4-chlorophenyl dichlorophosphate, 4-chlorophenylphosphorodichloridate, phosphorodichloridic acid, 4-chlorophenyl ester, 4-chlorophenylphosphoryl dichloride, acmc-1bjgo, cczmqygsxwzfki-uhfffaoysa, 4-chlorophenyl dichlorophosphinate, 4-chlorophenyl dichloridophosphate #
PubChem CID 69879
IUPAC Name 1-chloro-4-dichlorophosphoryloxybenzene
SMILES C1=CC(=CC=C1OP(=O)(Cl)Cl)Cl

Specifications

Density 1.508
Boiling Point 113°C to 115°C (0.9 mmHg)
Flash Point 106°C (222°F)
Odor Acrid
Refractive Index 1.5398
Quantity 25 g
UN Number UN3265
Beilstein 910754
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 245.43
Percent Purity ≥98%
Chemical Name or Material 4-Chlorophenyl phosphorodichloridate
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 212-254-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only