missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenyl Chlorothionoformate 98.0+%, TCI America™

Catalog No. C15885G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15885G 5 g
C158825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15885G Supplier TCI America Supplier No. C15885G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 937-64-4
Molecular Formula C7H4Cl2OS
Molecular Weight (g/mol) 207.068
MDL Number MFCD00014465
InChI Key BQIABQCJXBELMT-UHFFFAOYSA-N
Synonym 4-Chlorophenyl Chlorothioformate, Chlorothioformic Acid O-(4-Chlorophenyl) Ester
PubChem CID 70306
IUPAC Name O-(4-chlorophenyl) chloromethanethioate
SMILES C1=CC(=CC=C1OC(=S)Cl)Cl

Specifications

Color Yellow
Boiling Point 131°C
Quantity 5 g
UN Number 3265
Formula Weight 207.07
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 4-Chlorophenyl Chlorothionoformate