CAS RN 937-64-4
IUPAC Name:
4-chlorophenyl chloromethanethioate
Synonyms:
4-chlorophenyl chloromethanethioate
Molecular Weight (g/mol):
207.07
Molecular Formula:
C7H4Cl2OS
InChi Key:
BQIABQCJXBELMT-UHFFFAOYSA-N
SMILES:
ClC(=S)OC1=CC=C(Cl)C=C1
1
–
1
of
1
results
1
4-Chlorophenyl Chlorothionoformate 98.0+%, TCI America™
CAS: 937-64-4 Molecular Formula: C7H4Cl2OS Molecular Weight (g/mol): 207.068 MDL Number: MFCD00014465 InChI Key: BQIABQCJXBELMT-UHFFFAOYSA-N Synonym: 4-Chlorophenyl Chlorothioformate, Chlorothioformic Acid O-(4-Chlorophenyl) Ester PubChem CID: 70306 IUPAC Name: O-(4-chlorophenyl) chloromethanethioate SMILES: C1=CC(=CC=C1OC(=S)Cl)Cl