missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorophenacyl Bromide 98.0+%, TCI America™
Supplier: TCI America B0568100G
Specifications
| 4-Chlorophenacyl Bromide | |
| 99°C | |
| C8H6BrClO | |
| 100 g | |
| 2-bromo-4'-chloroacetophenone, 2-bromo-1-4-chlorophenyl ethanone, 4-chlorophenacyl bromide, p-chlorophenacyl bromide, 2-bromo-1-4-chlorophenyl ethan-1-one, ethanone, 2-bromo-1-4-chlorophenyl, 4'-chlorophenacyl bromide, 2-bromo-p-chloroacetophenone, p-bromoacetyl chlorobenzene, 4'-chloro-2-bromoacetophenone | |
| C1=CC(=CC=C1C(=O)CBr)Cl | |
| 233.489 | |
| 233.49 | |
| Crystalline Powder |
| 536-38-9 | |
| White-Yellow | |
| MFCD00000625 | |
| 3261 | |
| FLAYZKKEOIAALB-UHFFFAOYSA-N | |
| 2-bromo-1-(4-chlorophenyl)ethanone | |
| 68303 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 536-38-9 | |
| 233.489 | |
| FLAYZKKEOIAALB-UHFFFAOYSA-N | |
| 68303 | |
| C1=CC(=CC=C1C(=O)CBr)Cl |
| C8H6BrClO | |
| MFCD00000625 | |
| 2-bromo-4'-chloroacetophenone, 2-bromo-1-4-chlorophenyl ethanone, 4-chlorophenacyl bromide, p-chlorophenacyl bromide, 2-bromo-1-4-chlorophenyl ethan-1-one, ethanone, 2-bromo-1-4-chlorophenyl, 4'-chlorophenacyl bromide, 2-bromo-p-chloroacetophenone, p-bromoacetyl chlorobenzene, 4'-chloro-2-bromoacetophenone | |
| 2-bromo-1-(4-chlorophenyl)ethanone |
Safety and Handling
EINECSNumber : (3)-2934
RTECSNumber : AM5978800
TSCA : Yes