accessibility menu, dialog, popup

UserName

ClOBr

CAS RN 536-38-9

IUPAC Name: 2-bromo-1-(4-chlorophenyl)ethan-1-one
Synonyms: 2-bromo-1-(4-chlorophenyl)ethanone
Molecular Weight (g/mol): 233.49
Molecular Formula: C8H6BrClO
InChi Key: FLAYZKKEOIAALB-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C(=O)CBr
Molecular Weight (g/mol) 
  • (6)
  • (2)
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (3)
  • (3)
  • (2)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (6)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)