missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenacyl Bromide 98.0+%, TCI America™

Catalog No. p-7118531
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B056825G 25 g
B0568100G 100 g
B0568500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B056825G Supplier TCI America Supplier No. B056825G

Chemical Identifiers

CAS 536-38-9
Molecular Formula C8H6BrClO
Molecular Weight (g/mol) 233.489
MDL Number MFCD00000625
InChI Key FLAYZKKEOIAALB-UHFFFAOYSA-N
Synonym 2-bromo-4'-chloroacetophenone, 2-bromo-1-4-chlorophenyl ethanone, 4-chlorophenacyl bromide, p-chlorophenacyl bromide, 2-bromo-1-4-chlorophenyl ethan-1-one, ethanone, 2-bromo-1-4-chlorophenyl, 4'-chlorophenacyl bromide, 2-bromo-p-chloroacetophenone, p-bromoacetyl chlorobenzene, 4'-chloro-2-bromoacetophenone
PubChem CID 68303
IUPAC Name 2-bromo-1-(4-chlorophenyl)ethanone
SMILES C1=CC(=CC=C1C(=O)CBr)Cl

Specifications

Melting Point 99°C
Color White-Yellow
Quantity 25 g
UN Number 3261
Formula Weight 233.49
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chlorophenacyl Bromide
EINECSNumber (3)-2934
RTECSNumber AM5978800
TSCA Yes