missing translation for 'onlineSavingsMsg'
Learn More

4-Chloronitrobenzene 98.0+%, TCI America™

Supplier:  TCI America C022225G

 View more versions of this product

Catalog No. C022225G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-Chloronitrobenzene
84°C
C6H4ClNO2
25 g
4-chloronitrobenzene, p-chloronitrobenzene, p-nitrochlorobenzene, benzene, 1-chloro-4-nitro, 4-nitrochlorobenzene, pncb, 4-chloro-1-nitrobenzene, p-nitrophenyl chloride, p-nitrochlorobenzol, p-nitroclorobenzene
C1=CC(=CC=C1[N+](=O)[O-])Cl
157.553
CHEBI:34399
≥98.0% (GC)
100-00-5
242°C
MFCD00007285
1578
CZGCEKJOLUNIFY-UHFFFAOYSA-N
1-chloro-4-nitrobenzene
7474
157.55
Crystalline Powder

Chemical Identifiers

100-00-5
157.553
CZGCEKJOLUNIFY-UHFFFAOYSA-N
7474
1-chloro-4-nitrobenzene
C6H4ClNO2
MFCD00007285
4-chloronitrobenzene, p-chloronitrobenzene, p-nitrochlorobenzene, benzene, 1-chloro-4-nitro, 4-nitrochlorobenzene, pncb, 4-chloro-1-nitrobenzene, p-nitrophenyl chloride, p-nitrochlorobenzol, p-nitroclorobenzene
CHEBI:34399
C1=CC(=CC=C1[N+](=O)[O-])Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0442

RTECSNumber : CZ1050000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions